Skip to Content
MilliporeSigma
3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt for enzymic, spectrophotometric determination of H2O2, ≥98.0%

3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt

3-(N-Ethyl-3-methylanilino)-2-hydroxypropanesulfonic acid sodium salt for enzymic, spectrophotometric determination of H2O2, ≥98.0%
Synonym(s):
3-(N-Ethyl-m-toluidino)-2-hydroxypropanesulfonic acid sodium salt, N-Ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine sodium salt, Sodium 3-(N-ethyl-3-methylanilino)-2-hydroxypropanesulfonate, Sodium N-ethyl-N-(2-hydroxy-3-sulfopropyl)-m-toluidine, TOOS
Empirical Formula (Hill Notation):
C12H18NNaO4S
CAS Number:
Molecular Weight:
295.33

Products

Formula Weight
Physical Form
Purity
Format
Quality Segment
Application
Special Grade
Technique