Skip to Content
MilliporeSigma
3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥99.0% (HPLC)

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate

3-Methyl-2-benzothiazolinone hydrazone hydrochloride monohydrate ≥99.0% (HPLC)
Synonym(s):
2-Hydrazono-3-methylbenzothiazoline hydrochloride
Empirical Formula (Hill Notation):
C8H9N3S · HCl · H2O
CAS Number:
Molecular Weight:
233.72
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1