Skip to Content
MilliporeSigma
3-Methoxyphenylacetonitrile 99%

3-Methoxyphenylacetonitrile

3-Methoxyphenylacetonitrile 99%
Synonym(s):
3-Methoxybenzyl cyanide
Linear Formula:
CH3OC6H4CH2CN
CAS Number:
Molecular Weight:
147.17
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing