Skip to Content
MilliporeSigma
3-Hydroxybenzylhydrazine dihydrochloride ≥98.0% (HPLC)

3-Hydroxybenzylhydrazine dihydrochloride

3-Hydroxybenzylhydrazine dihydrochloride ≥98.0% (HPLC)
Synonym(s):
α-Hydrazino-m-cresol dihydrochloride, 3-(Hydrazinomethyl)phenol dihydrochloride, NSD-1015
Linear Formula:
HOC6H4CH2NHNH2 · 2HCl
CAS Number:
Molecular Weight:
211.09
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Research Area
Showing 1-1 of 1
Compare
Product Number
Description
Pricing