Skip to Content
MilliporeSigma
3-Formylphenylboronic acid ≥95%

3-Formylphenylboronic acid

3-Formylphenylboronic acid ≥95%
Synonym(s):
(3-Formylbenzene)boronic acid, 3-(Dihydroxyboryl)benzaldehyde, 3-Boronobenzaldehyde, m-Formylphenylboronic acid, m-formyl-benzeneboronic acid
Linear Formula:
HCOC6H4B(OH)2
CAS Number:
Molecular Weight:
149.94

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing