Skip to Content
MilliporeSigma
3-Fluoro-2-formylphenylboronic acid ≥95%

3-Fluoro-2-formylphenylboronic acid

3-Fluoro-2-formylphenylboronic acid ≥95%
Empirical Formula (Hill Notation):
C7H6BFO3
CAS Number:
Molecular Weight:
167.93

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing