Skip to Content
MilliporeSigma
3-Dehydroshikimic acid ≥95.0% (HPLC)

3-Dehydroshikimic acid

3-Dehydroshikimic acid ≥95.0% (HPLC)
Synonym(s):
(4S,5R)-(−)-4,5-Dihydroxy-3-oxo-1-cyclohexene-1-carboxylic acid, 3-DHS, 5-Dehydroshikimic acid
Empirical Formula (Hill Notation):
C7H8O5
CAS Number:
Molecular Weight:
172.14

Products

Brand
Color
Formula Weight
Physical Form
Purity
Showing 1-1 of 1
Compare
Product Number
Description
Pricing