Skip to Content
MilliporeSigma
(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution 60 wt. % in H2O

(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution

(3-Chloro-2-hydroxypropyl)trimethylammonium chloride solution 60 wt. % in H2O
Linear Formula:
ClCH2CH(OH)CH2N(Cl)(CH3)3
CAS Number:
Molecular Weight:
188.10

Products

Brand
Formula Weight
Physical Form
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing