Skip to Content
MilliporeSigma
3-Bromothiophene-2-carboxaldehyde 96%

3-Bromothiophene-2-carboxaldehyde

3-Bromothiophene-2-carboxaldehyde 96%
Empirical Formula (Hill Notation):
C5H3BrOS
CAS Number:
Molecular Weight:
191.05

Products

Product Category
Brand
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing