Skip to Content
MilliporeSigma
3-Bromocyclohexene technical grade, 90%

3-Bromocyclohexene

3-Bromocyclohexene technical grade, 90%
Synonym(s):
2-Cyclohexen-1-yl bromide
Empirical Formula (Hill Notation):
C6H9Br
CAS Number:
Molecular Weight:
161.04

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing