Skip to Content
MilliporeSigma
3-Bromoadamantane-1-carboxylic acid 97%

3-Bromoadamantane-1-carboxylic acid

3-Bromoadamantane-1-carboxylic acid 97%
Empirical Formula (Hill Notation):
C11H15BrO2
CAS Number:
Molecular Weight:
259.14

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing