Skip to Content
MilliporeSigma
3-Bromo-5-fluoropyridine-2-carboxylic acid 97%

3-Bromo-5-fluoropyridine-2-carboxylic acid

3-Bromo-5-fluoropyridine-2-carboxylic acid 97%
Empirical Formula (Hill Notation):
C6H3BrFNO2
CAS Number:
Molecular Weight:
220.00

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing