Skip to Content
MilliporeSigma
3′-Bromo-2,2,2-trifluoroacetophenone ≥97%

3′-Bromo-2,2,2-trifluoroacetophenone

3′-Bromo-2,2,2-trifluoroacetophenone ≥97%
Synonym(s):
3-Bromo-α,α,α-trifluoroacetophenone
Empirical Formula (Hill Notation):
C8H4BrF3O
CAS Number:
Molecular Weight:
253.02

Products

Product Category
Brand
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing