Skip to Content
MilliporeSigma
(3aR)-(+)-Sclareolide natural, 97%, FG

(3aR)-(+)-Sclareolide

(3aR)-(+)-Sclareolide natural, 97%, FG
Empirical Formula (Hill Notation):
C16H26O2
CAS Number:
Molecular Weight:
250.38
EC Number:

Products

Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Melting Point (°C)
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing