Skip to Content
MilliporeSigma
3-Aminopyrazole-4-carbonitrile 97%

3-Aminopyrazole-4-carbonitrile

3-Aminopyrazole-4-carbonitrile 97%
Synonym(s):
3-Amino-4-cyanopyrazole
Empirical Formula (Hill Notation):
C4H4N4
CAS Number:
Molecular Weight:
108.10
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing