Skip to Content
MilliporeSigma
3-Aminopropylphosphonic acid 98%

3-Aminopropylphosphonic acid

3-Aminopropylphosphonic acid 98%
Linear Formula:
H2N(CH2)3P(O)(OH)2
CAS Number:
Molecular Weight:
139.09

Products

Product Category
Brand
Formula Weight
Gene Alias
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing