Skip to Content
MilliporeSigma
3,5-Dichloroanisole 98%

3,5-Dichloroanisole

3,5-Dichloroanisole 98%
Synonym(s):
1,3-Dichloro-5-methoxybenzene
Linear Formula:
Cl2C6H3OCH3
CAS Number:
Molecular Weight:
177.03
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing