Skip to Content
MilliporeSigma
3,5-Bis(trifluoromethyl)phenylboronic acid ≥95%

3,5-Bis(trifluoromethyl)phenylboronic acid

3,5-Bis(trifluoromethyl)phenylboronic acid ≥95%
Synonym(s):
3,5-Di(trifluoromethyl)benzeneboronic acid
Linear Formula:
(CF3)2C6H3B(OH)2
CAS Number:
Molecular Weight:
257.93

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing