Skip to Content
MilliporeSigma
3,4-Di-O-acetyl-6-deoxy-L-glucal 98%

3,4-Di-O-acetyl-6-deoxy-L-glucal

3,4-Di-O-acetyl-6-deoxy-L-glucal 98%
Synonym(s):
Di-O-acetyl-L-rhamnal
Empirical Formula (Hill Notation):
C10H14O5
CAS Number:
Molecular Weight:
214.22
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Optical Isomer
Showing 1-1 of 1
Compare
Product Number
Description
Pricing