Skip to Content
MilliporeSigma
3,4-Dihydroxybenzylamine hydrobromide 98%

3,4-Dihydroxybenzylamine hydrobromide

3,4-Dihydroxybenzylamine hydrobromide 98%
Synonym(s):
4-(Aminomethyl)catechol hydrobromide, DHBA hydrobromide
Linear Formula:
(HO)2C6H3CH2NH2 · HBr
CAS Number:
Molecular Weight:
220.06
EC Number:

Products

Product Category
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing