Skip to Content
MilliporeSigma
3,4,5-Trimethoxyphenylacetonitrile 97%

3,4,5-Trimethoxyphenylacetonitrile

3,4,5-Trimethoxyphenylacetonitrile 97%
Synonym(s):
3,4,5-Trimethoxybenzyl cyanide
Linear Formula:
(CH3O)3C6H2CH2CN
CAS Number:
Molecular Weight:
207.23
EC Number:

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing