3,3′-Di-O-methylellagic acid 4′-xylopyranoside
Synonym(s):
3,3′-Di-O-methyl ellagic acid 4′-O-β-D-xylopyranoside, 2-Hydroxy-3,8-dimethoxy-7-(β-D-xylopyranosyloxy)-[1]benzopyrano[5,4,3-cde][1]benzopyran-5,10-dione, 3,3′-Di-O-methyl-4-O-((β-D-xylopyranosyl)ellagic acid, 3-O-Methylducheside A
Empirical Formula (Hill Notation):
C21H18O12
CAS Number:
Molecular Weight:
462.36
Products
Formula Weight
Manufacturer
Physical Form
Product Line
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing