Skip to Content
MilliporeSigma
3,3′-Dioctadecyloxacarbocyanine perchlorate

3,3′-Dioctadecyloxacarbocyanine perchlorate

3,3′-Dioctadecyloxacarbocyanine perchlorate
Synonym(s):
DiOC18(3), ‘DiO’
Empirical Formula (Hill Notation):
C53H85ClN2O6
CAS Number:
Molecular Weight:
881.70

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Application
Showing 1-1 of 1
Compare
Product Number
Description
Pricing