Skip to Content
MilliporeSigma
3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol 97%

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol

3,3,4,4,5,5,6,6,7,7,8,8,8-Tridecafluoro-1-octanethiol 97%
Synonym(s):
1H,1H,2H,2H-Perfluoro-1-octanethiol, 1H,1H,2H,2H-Perfluorooctyl mercaptan
Linear Formula:
CF3(CF2)5CH2CH2SH
CAS Number:
Molecular Weight:
380.17

Products

Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing