Skip to Content
MilliporeSigma
2-(Trimethylsilyl)ethanethiol 95%

2-(Trimethylsilyl)ethanethiol

2-(Trimethylsilyl)ethanethiol 95%
Synonym(s):
(2-Mercaptoethyl)trimethylsilane, BEST
Linear Formula:
(CH3)3SiCH2CH2SH
CAS Number:
Molecular Weight:
134.32

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing