Skip to Content
MilliporeSigma
2-Thienylboronic acid ≥95.0%

2-Thienylboronic acid

2-Thienylboronic acid ≥95.0%
Synonym(s):
2-Thienylboric acid, 2-Thienylboronic acid, Thien-5-ylboronic acid, Thiophene-2-boronic acid
Empirical Formula (Hill Notation):
C4H5BO2S
CAS Number:
Molecular Weight:
127.96

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing