Skip to Content
MilliporeSigma
(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone 97%

(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone

(2S,5S)-(−)-2-tert-Butyl-3-methyl-5-benzyl-4-imidazolidinone 97%
Synonym(s):
(2S,5S)-2-tert-Butyl-3-methyl-5-phenylmethyl-4-imidazolidinone, (2S,5S)-5-Benzyl-2-tert-butyl-3-methyl-4-imidazolidinone
Empirical Formula (Hill Notation):
C15H22N2O
CAS Number:
Molecular Weight:
246.35

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing