Skip to Content
MilliporeSigma
(2R)-(−)-Glycidyl tosylate 98%

(2R)-(−)-Glycidyl tosylate

(2R)-(−)-Glycidyl tosylate 98%
Synonym(s):
(R)-(−)-Glycidyl p-toluenesulfonate, (R)-(−)-Glycidyl tosylate, (R)-(−)-Oxirane-2-methanol p-toluenesulfonate salt
Empirical Formula (Hill Notation):
C10H12O4S
CAS Number:
Molecular Weight:
228.26

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing