Skip to Content
MilliporeSigma
2-(Pyridin-2-yl)isopropyl amine 95% (GC)

2-(Pyridin-2-yl)isopropyl amine

2-(Pyridin-2-yl)isopropyl amine 95% (GC)
Empirical Formula (Hill Notation):
C8H12N2
CAS Number:
Molecular Weight:
136.19

Products

Formula Weight
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing