Skip to Content
MilliporeSigma
2-Propanol-d8 99.5 atom % D

2-Propanol-d8

2-Propanol-d8 99.5 atom % D
Synonym(s):
Isopropanol-d8, Octadeuteroisopropanol
Linear Formula:
CD3CD(OD)CD3
CAS Number:
Molecular Weight:
68.14
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Isotope
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing