Skip to Content
MilliporeSigma
2-O-a-D-Glucopyranosyl-L-ascorbic Acid ≥98% (HPLC)

2-O-a-D-Glucopyranosyl-L-ascorbic Acid

2-O-a-D-Glucopyranosyl-L-ascorbic Acid ≥98% (HPLC)
Synonym(s):
Ascorbyl Glucoside
Empirical Formula (Hill Notation):
C12H18O11
CAS Number:
Molecular Weight:
338.26

Products

Product Category
Brand
Color
Formula Weight
Physical Form
Purity
Quality Segment
Technique
Showing 1-1 of 1
Compare
Product Number
Description
Pricing