
2-Nitrobenzenesulfonyl chloride

Synonym(s):
2-Nitrobenzenesulfonyl chloride
Linear Formula:
O2NC6H4SO2Cl
CAS Number:
Molecular Weight:
221.62
Products
Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Technique
Showing 1-2 of 2
Compare
Product Number
Description
Pricing