Skip to Content
MilliporeSigma
2-Methyl-3-heptanone 99%

2-Methyl-3-heptanone

2-Methyl-3-heptanone 99%
Synonym(s):
Butyl isopropyl ketone
Linear Formula:
CH3(CH2)3COCH(CH3)2
CAS Number:
Molecular Weight:
128.21
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing