Skip to Content
MilliporeSigma
2-Methyl-3-buten-2-ol 98%

2-Methyl-3-buten-2-ol

2-Methyl-3-buten-2-ol 98%
Synonym(s):
1,1-Dimethylallyl alcohol, 3-Hydroxy-3-methyl-1-butene, 2-Methyl-3-buten-2-ol
Linear Formula:
CH2=CHC(CH3)2OH
CAS Number:
Molecular Weight:
86.13

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing