2-Methyl-2-phenylpropylmagnesium chloride solution
Linear Formula:
C6H5C(CH3)2CH2MgCl
CAS Number:
Molecular Weight:
192.97
Products
Brand
Boiling Point (°C)
Formula Weight
Markush Class
Markush Group
Reaction Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing