Skip to Content
MilliporeSigma
2-Methoxyphenylboronic acid 95%

2-Methoxyphenylboronic acid

2-Methoxyphenylboronic acid 95%
Synonym(s):
(2-Methoxyphenyl)boric acid, 2-Methoxybenzeneboronic acid, o-Anisylboronic acid, o-Methoxybenzeneboronic acid, o-Methoxyphenylboronic acid
Linear Formula:
CH3OC6H4B(OH)2
CAS Number:
Molecular Weight:
151.96

Products

Formula Weight
Melting Point (°C)
Physical Form
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing