Skip to Content
MilliporeSigma
2-Methoxyphenylacetic acid 98%

2-Methoxyphenylacetic acid

2-Methoxyphenylacetic acid 98%
Synonym(s):
2-Methoxyphenylacetic acid
Linear Formula:
2-(CH3O)C6H4CH2COOH
CAS Number:
Molecular Weight:
166.17

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2
Compare
Product Number
Description
Pricing