Skip to Content
MilliporeSigma
2-Mercaptopyridine N-oxide sodium salt 95%

2-Mercaptopyridine N-oxide sodium salt

2-Mercaptopyridine N-oxide sodium salt 95%
Synonym(s):
1-Hydroxy-2-pyridinethione sodium salt, 2-Mercaptopyridine-1-oxide sodium salt, 2-Pyridinethiol-1-oxide sodium salt, Pyrithione sodium salt
Empirical Formula (Hill Notation):
C5H4NNaOS
CAS Number:
Molecular Weight:
149.15

Products

Product Category
Brand
Biological Source
Color
Formula Weight
Purity
Activity Spectrum
Markush Class
Markush Group
Mode of Action
Showing 1-2 of 2
Compare
Product Number
Description
Pricing