Skip to Content
MilliporeSigma
2-(Isopropylamino)ethanol 70%

2-(Isopropylamino)ethanol

2-(Isopropylamino)ethanol 70%
Synonym(s):
N-Isopropylethanolamine
Linear Formula:
(CH3)2CHNHCH2CH2OH
CAS Number:
Molecular Weight:
103.16

Products

Product Category
Boiling Point (°C)
Formula Weight
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing