Skip to Content
MilliporeSigma
2-Isopropyl-5-methyl-2-hexenal ≥95%, stabilized, FG

2-Isopropyl-5-methyl-2-hexenal

2-Isopropyl-5-methyl-2-hexenal ≥95%, stabilized, FG
Linear Formula:
(CH3)2CHCH2CH=C[CH(CH3)2]CHO
CAS Number:
Molecular Weight:
154.25
EC Number:

Products

Product Category
Biological Source
Boiling Point (°C)
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing