Skip to Content
MilliporeSigma
2-Isocyanatoethyl methacrylate contains ≤0.1% BHT as inhibitor, 98%

2-Isocyanatoethyl methacrylate

2-Isocyanatoethyl methacrylate contains ≤0.1% BHT as inhibitor, 98%
Synonym(s):
2-(Methacryloyloxy)ethyl isocyanate, IEM
Linear Formula:
H2C=C(CH3)CO2CH2CH2NCO
CAS Number:
Molecular Weight:
155.15
EC Number:

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing