Skip to Content
MilliporeSigma
2-Isobutylthiazole 99%

2-Isobutylthiazole

2-Isobutylthiazole 99%
Empirical Formula (Hill Notation):
C7H11NS
CAS Number:
Molecular Weight:
141.23
EC Number:

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Agency
Quality Segment
Application
Markush Class
Markush Group
Organoleptic Class
Special Grade
Showing 1-2 of 2
Compare
Product Number
Description
Pricing