Skip to Content
MilliporeSigma
2-Imidazolecarboxaldehyde 97%

2-Imidazolecarboxaldehyde

2-Imidazolecarboxaldehyde 97%
Empirical Formula (Hill Notation):
C4H4N2O
CAS Number:
Molecular Weight:
96.09

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing