Skip to Content
MilliporeSigma
2-Hydroxypropyl methacrylamide 99% (GC)

2-Hydroxypropyl methacrylamide

2-Hydroxypropyl methacrylamide 99% (GC)
Synonym(s):
N-(2-Hydroxypropyl)-2-methyl-prop-2-enamide
Empirical Formula (Hill Notation):
C7H13NO2
CAS Number:
Molecular Weight:
143.18

Products

Brand
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Monomer Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing