Skip to Content
MilliporeSigma
2-Hydroxymethyl-18-crown-6 95%

2-Hydroxymethyl-18-crown-6

2-Hydroxymethyl-18-crown-6 95%
Synonym(s):
1,4,7,10,13,16-Hexaoxacyclooctadecane-2-methanol, 18-Crown-6-methanol
Empirical Formula (Hill Notation):
C13H26O7
CAS Number:
Molecular Weight:
294.34
EC Number:

Products

Brand
Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing