Skip to Content
MilliporeSigma
2-Hydroxymethyl-15-crown-5 95%

2-Hydroxymethyl-15-crown-5

2-Hydroxymethyl-15-crown-5 95%
Synonym(s):
1,4,7,10,13-Pentaoxacyclopentadecane-2-methanol
Empirical Formula (Hill Notation):
C11H22O6
CAS Number:
Molecular Weight:
250.29
EC Number:

Products

Boiling Point (°C)
Formula Weight
Physical Form
Purity
Functional Group
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing