Skip to Content
MilliporeSigma
2-Hexyl-2-decenal mixture of cis and trans, ≥95%

2-Hexyl-2-decenal

2-Hexyl-2-decenal mixture of cis and trans, ≥95%
Empirical Formula (Hill Notation):
C16H30O
CAS Number:
Molecular Weight:
238.41

Products

Product Category
Brand
Sustainability
Biological Source
Feature
Formula Weight
Purity
Agency
Quality Segment
Application
Organoleptic Class
Special Grade