Skip to Content
MilliporeSigma
2-Ethylhexyl acetate 99%

2-Ethylhexyl acetate

2-Ethylhexyl acetate 99%
Synonym(s):
2-Ethylhexyl acetate
Linear Formula:
CH3COOCH2CH(C2H5)(CH2)3CH3
CAS Number:
Molecular Weight:
172.26

Products

Product Category
Brand
Biological Source
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Application
Organoleptic Class
Special Grade
Showing 1-3 of 3
Compare
Product Number
Description
Pricing