Skip to Content
MilliporeSigma
2-Ethyl-9,10-dimethoxyanthracene 97%

2-Ethyl-9,10-dimethoxyanthracene

2-Ethyl-9,10-dimethoxyanthracene 97%
Empirical Formula (Hill Notation):
C18H18O2
CAS Number:
Molecular Weight:
266.33

Products

Product Category
Brand
Formula Weight
Melting Point (°C)
Purity
Markush Class
Markush Group
Showing 1-1 of 1
Compare
Product Number
Description
Pricing