Skip to Content
MilliporeSigma
2-Ethyl-1,3-hexanediol 97%, Mixture of isomers

2-Ethyl-1,3-hexanediol

2-Ethyl-1,3-hexanediol 97%, Mixture of isomers
Synonym(s):
Ethylhexylene glycol, 2-Ethyl-1,3-hexanediol
Linear Formula:
CH3CH2CH2CH(OH)CH(C2H5)CH2OH
CAS Number:
Molecular Weight:
146.23

Products

Product Category
Brand
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Markush Class
Markush Group
Showing 1-2 of 2